Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lecithin
Lecithin (from egg yolk)
Lecithin (from egg yolk)
Lecithin (from egg yolk)
Catalog # 80053-676
Supplier:
MilliporeSigma
CAS Number:
SDS
Certificates
Expand image
Lecithin (from egg yolk)
Catalog # 80053-676
Supplier:
MilliporeSigma
Supplier Number:
524617-250MG
CAS Number:
SDS
Certificates
Specifications
Size:
250 mg
Cat. No.:
80053-676
Supplier no.:
524617-250MG
Specifications
About this item
Product Details
Documentation
:
CAS Number:
8002-43-5
MDL Number:
MFCD00147406
Product Details
Documentation