ContactUsLinkComponent
Brady® SPC® Universal Granular Absorbent, Brady Worldwide
Brady® SPC® Universal Granular Absorbent, Brady Worldwide
Catalog # 76193-316
Supplier:  Brady Worldwide
undefined
Brady® SPC® Universal Granular Absorbent, Brady Worldwide
Catalog # 76193-316
Supplier:  Brady Worldwide
Supplier Number:  DZ-100
Specifications
  • Description:
    Granular Absorbent
  • Size:
    40×40 lb.
  • Environmentally Preferable:
  • Cat. No.:
    76193-316
  • Packaging:
    BAGA
  • Absorbency:
    8.5 gal.
Specifications
About this item
Universal Granular Absorbent is a loose, multi-purpose absorbent ideal for liquid spill clean-up, lab packing, and stabilization of free liquids. Highly versatile absorbents pick up water-based, petroleum-based, and non-aggressive chemical fluids. Corn Cob Granular is perfect for aircraft maintenance, machine shops, food processing, filling lines, and bulk liquid transfer stations. Not for use with aggressive fluids.
  • Absorbency capacity: 8.5 gallons
  • Packaged in a 40 lb. bag