ContactUsLinkComponent
BD BBL™ MacConkey II Agar with MUG and Trypticase™ Soy Agar with 5% Sheep Blood (TSA II), BD Diagnostics
Catalog # 90006-558
Supplier:  Avantor
CAS Number:  
BD BBL™ MacConkey II Agar with MUG and Trypticase™ Soy Agar with 5% Sheep Blood (TSA II), BD Diagnostics
Catalog # 90006-558
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Pack type:
    Plate
  • Description:
    BBL™ Trypticase™ Soy Agar with 5% Sheep Blood (TSA II) and BBL™ MacConkey II Agar with MUG–I Plate™
  • Size:
    20/pack
  • Environmentally Preferable:
  • Cat. No.:
    90006-558
  • Supplier no.:
    221949
Specifications
About this item
The Prepared Plated Media of BBL™ Trypticase™ Soy Agar with 5% Sheep Blood is used for the growth of fastidious organisms and for the visualization of hemolytic reactions and MacConkey II Agar with MUG is used for the presumptive identification of Escherichia coli.