Order Entry
United States
ContactUsLinkComponent
Intracellular Staining Permeabilization Wash Buffer (10X), Biolegend
Intracellular Staining Permeabilization Wash Buffer (10X), Biolegend
Catalog # 421002-BL
Supplier:  Biolegend
CAS Number:  
Intracellular Staining Permeabilization Wash Buffer (10X), Biolegend
Catalog # 421002-BL
Supplier:  Biolegend
Supplier Number:  421002
CAS Number:  

Specifications

  • Description:
    Intracellular Staining Permeabilization Wash Buffer (10X)
  • Size:
    100 mL
  • Storage Temperature:
    2 – 8 °C
  • Cat. No.:
    421002-BL
  • Supplier no.:
    421002

Specifications

About this item

Intracellular staining permeabilization wash buffer is useful for intracellular staining procedures, e.g., in preparation of cells for staining intracellular cytokines or other proteins. Intracellular staining permeabilization wash buffer is used to permeabilize cells following fixation with intracellular staining fixation buffer.

Intracellular staining permeabilization wash buffer has been formulated to have minimal effects on cells, reduce non-specific staining and enhance the signal to noise ratio. It can be used for antibody dilutions and cell washing during intracellular staining.

The Intracellular staining perm wash buffer solution should be stored between 2 °C and 8 °C. Do not freeze.

Delivery: It is supplied as a 10X solution and should be diluted in deionized water prior to use.