Order Entry
United States
ContactUsLinkComponent
 
pNLCoI4[luc2-P2A-NlucP/PGK/Hygro] Vector, 20 µg, Promega
Catalog # PAN1491
CAS Number:  
undefined
pNLCoI4[luc2-P2A-NlucP/PGK/Hygro] Vector, 20 µg, Promega
Catalog # PAN1491
Supplier Number:  N1491
CAS Number:  

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAN1491
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    N1491

Specifications

About this item

The pNLCoI Vectors comprise a second-generation coincidence reporter vector system that allow expression of both firefly luciferase (luc2) and NanoLuc Luciferase fused to a PEST destabilization domain (NlucP) from the same mRNA transcript.

  • Vectors That Express Both Firefly and NanoLuc(R) Luciferases from the Same Transcript
  • Firefly and NanoLuc(R) luciferase have dissimilar compound interference profiles for better identifying more false-positives versus individually
  • Using two different transcriptional reporters reduces false hit rates and increases the identification of true biological hits
  • luc2 and NlucP provide a bright reporter combination compatible with low-copy-number and plate scale-up, and provide greater signal-to-background ratios

Luciferase-based reporter-gene assays remain a useful and powerful method of high-throughput compound screening. However, false hits that result from direct interaction of compounds with the luciferase reporter can result in unnecessary follow-up efforts. The pNLCoI Vectors comprise a second-generation coincidence reporter vector system that allow expression of both firefly luciferase (luc2) and NanoLuc Luciferase fused to a PEST destabilization domain (NlucP) from the same mRNA transcript. The stoichiometric expression of both luciferases is achieved by use of the P2A sequence from porcine teschovirus-1, which promotes a ribosomal skip and expression of the two unfused enzymes with distinct compound interaction profiles. When used in high-throughput compound screening, false hits caused by direct interaction with one or the other luciferases can be distinguished from true hits that show a similar response for both, reducing workload associated with follow-up screens. The pNLCoI Vectors are designed for use with the Nano-Glo Dual-Luciferase Reporter (NanoDLR) Assay System, which allows sequential detection of firefly and NanoLuc Luciferase in activity in the same sample. Both reagents provide stable glow-type luminescence signals with half-lives of approximately two hours allowing batch processing of samples and amenable to assays or screens in 96-, 384- or 1,536-well plate formats. Potent inhibition of firefly luciferase coupled with the high-intensity luminescence of NanoLuc luciferase maximizes sensitivity for detection of both reporters.