Order Entry
United States
ContactUsLinkComponent
pFN31A Nluc CMV-Hygro Flexi Vector, 20 µg, Promega
pFN31A Nluc CMV-Hygro Flexi Vector, 20 µg, Promega
Catalog # PAN1311
CAS Number:  
undefined
pFN31A Nluc CMV-Hygro Flexi Vector, 20 µg, Promega
Catalog # PAN1311
Supplier Number:  N1311
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAN1311
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    N1311

Specifications

About this item

The pFN31 and pFC32 Vectors generate N- or C-terminal fusions of NanoLuc luciferase with your protein of interest. Use pFN31A/K Nluc CMV-Hygro Flexi Vectors for N-terminal fusions and pFC32A/K Nluc CMV-Hygro Flexi Vectors for C-terminal fusions.

  • Generate N- or C-Terminal Fusions to NanoLuc(R) Luciferase Reporter
  • Insert protein of interest using the Flexi(R) Vector System
  • Select options for N- or C-terminal NanoLuc(R) fusions and antibiotic resistance (ampicillin or kanamycin)
  • Bright NanoLuc(R) reporter enables endogenous expression levels of fusion proteins for biologically relevant results

The small size (19.1kDa) and extreme brightness (about 100-fold brighter than either firefly [Photinus pyralis] or Renilla reniformis) of NanoLuc luciferase (Nluc) make it an ideal protein fusion partner. NanoLuc fusion proteins can be used in a variety of applications including: reporters of protein stability, probes for bioluminescent cell imaging (BLI) or as the donor signal in bioluminescent resonance energy transfer (BRET) applications for protein:protein or protein:small-molecule interaction studies. The NanoLuc protein fusion vectors enable simple generation of N- or C-terminal fusions of NanoLuc luciferase with your protein of interest and are available. The pF Vector series generates N- or C-terminal Nluc fusion proteins using the Flexi Vector Cloning System—a directional cloning method based on two rare-cutting restriction enzymes, SgfI and PmeI, that provides a rapid, efficient and high-fidelity way to transfer protein-coding regions between a variety of Flexi Vectors without the need to resequence. Select the pFN31A and pFN31K Nluc CMV-Hygro Flexi Vectors for N-terminal protein fusions and pFC32A and pFC32K Nluc CMV-Hygro Flexi Vectors for C-terminal protein fusions. Also make NanoLuc protein fusions with the pNLF Vector series: Generate N- or C-terminal fusions to the full-length Nluc protein or attach secreted Nluc to the N-terminus of the protein of interest using traditional cloning with a multiple cloning site (MCS).