Order Entry
United States
ContactUsLinkComponent
pF3K WG (BYDV) Flexi Vector, 20 µg, Promega
pF3K WG (BYDV) Flexi Vector, 20 µg, Promega
Catalog # PAL5681
CAS Number:  
undefined
pF3K WG (BYDV) Flexi Vector, 20 µg, Promega
Catalog # PAL5681
Supplier Number:  L5681
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAL5681
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    L5681

Specifications

About this item

Designed for use with Wheat Germ extracts and contain sequences from the barley yellow dwarf virus (BYDV), an RNA plant virus, upstream and downstream of the protein coding region of interest.

  • Protein Expression Vectors Used with Wheat Germ Cell-Free Systems
  • Cloned inserts are in vitro translated in WGE systems
  • Adaptable to high-throughput formats for large screening projects
  • Transfer your cloned insert into other Flexi(R) Vectors for additional applications (e.g., bacterial or mammalian protein expression) as needed

The Flexi Vector System is a simple, yet powerful, directional cloning method for protein-coding sequences. It is based on two rare-cutting restriction enzymes, SgfI and PmeI, and provides a rapid, efficient and high-fidelity way to transfer protein-coding regions between a variety of Flexi Vectors without the need to resequence. The vectors are designed with special sequences for maximal cell-free protein expression in a specific system. The pF3A/K WG vectors were designed for use with Wheat Germ extracts and contain sequences from the barley yellow dwarf virus (BYDV), an RNA plant virus, upstream and downstream of the protein coding region of interest. The BYDV elements interact with each other, form a closed loop and act synergistically to stimulate translation in wheat germ extracts, bypassing mRNA cap and polyadenylation dependencies. Note: Flexi Vectors contain the lethal barnase gene to reduce background colonies without inserts during the subcloning procedure. Using the Flexi Vector Cloning System replaces the barnase gene with your insert. These vectors, as purchased, cannot be cultured in normal laboratory strains of E. coli without an insert.