Order Entry
United States
ContactUsLinkComponent
pFC20K HaloTag T7 SP6 Flexi Vector, 20 µg, Promega
pFC20K HaloTag T7 SP6 Flexi Vector, 20 µg, Promega
Catalog # PAG1691
Supplier:  Avantor
CAS Number:  
pFC20K HaloTag T7 SP6 Flexi Vector, 20 µg, Promega
Catalog # PAG1691
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAG1691
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    G1691

Specifications

About this item

These vectors are used for inducible expression of HaloTag fusion proteins in E. coli and cell-free systems using the T7 and SP6 RNA polymerase promoters. Vectors are part of the Flexi Cloning System.

  • HaloTag(R) Fusion Protein Vectors for Bacterial Expression
  • Express HaloTag(R) fusion proteins from T7 and SP6 promoters in E. coli and cell-free systems
  • HaloTag(R) technology for expression and purification of fusion proteins
  • Select options for N- or C-terminal HaloTag(R) fusions and antibiotic resistance

The following vectors are used for inducible expression of HaloTag fusion proteins in E. coli and cell-free systems using the T7 and SP6 RNA polymerase promoters. Expression levels depend highly on the nature of the protein, but in general the N-terminal HaloTag fusion protein can increase expression level, enhance refolding and boost solubility of the expressed protein. HaloTag vectors are supplied as Flexi System vectors. The Flexi Vector System is a simple, directional cloning method for protein-coding sequences. It is based on two rare-cutting restriction enzymes, SgfI and PmeI, and provides a rapid, efficient and high-fidelity way to transfer protein-coding regions between a variety of Flexi Vectors without the need to resequence. Direct transfers can only occur between two N-terminal tagged vectors or from an N-terminal to a C-terminal vector. pFN19A/K HaloTag T7 SP6 Flexi Vectors (Cat.# G1891, G1841) are designed for protein expression with an N-terminal HaloTag in T7 and SP6 cell-free expression systems. These vectors are optimized for cell-free expression systems. pFC20A/K HaloTag T7 SP6 Flexi Vectors (Cat.# G1681, G1691) are designed for protein expression with a C-terminal HaloTag in E. coli and SP6 cell-free expression systems. These vectors are optimized for cell-free expression systems. Note: Flexi Vectors contain the lethal barnase gene to reduce background colonies without inserts during the subcloning procedure. Using the Flexi Vector Cloning System replaces the barnase gene with your insert. These vectors, as purchased, cannot be cultured in normal laboratory strains of E. coli without an insert.