Order Entry
United States
ContactUsLinkComponent
pGL4.25[luc2CP/minP] Vector, 20 µg, Promega
pGL4.25[luc2CP/minP] Vector, 20 µg, Promega
Catalog # PAE8431
CAS Number:  
undefined
pGL4.25[luc2CP/minP] Vector, 20 µg, Promega
Catalog # PAE8431
Supplier Number:  E8431
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAE8431
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    E8431

Specifications

About this item

pGL4.23[luc2/minP], pGL4.24[luc2P/minP] and pGL4.25[luc2CP/minP] Vectors offer different firefly luciferase protein stabilities for expression in mammalian cells. Clone a response element of interest upstream of the minimal promoter and reporter gene.

  • Vectors Used for Cloning a Response Element Upstream of Firefly Luciferase Gene
  • Minimal promoter (minP) helps drive expression of luciferase when a response element is cloned upstream
  • Available with three varieties of engineered firefly luciferase genes: luc2, luc2P or luc2CP

Creating a cell line with an indicator of a functional signaling pathway is useful for deciphering the components in a signaling pathway. These tools are made by inserting multiple repeats of a response element upstream of a minimal promoter (minP). To construct pathway reporters, there are minimal promoter (minP) vectors with three varieties of engineered firefly luciferase genes: luc2, luc2P or luc2CP. The luc2 gene is engineered to remove most cryptic transcription factor binding sites and improve mammalian expression through codon optimization. The luc2P and luc2CP and RapidResponse genes are luc2 genes appended with degradation sequences to influence the cellular half-life of the luc2 gene. The RapidResponse genes respond more rapidly to stimuli but at the expense of signal intensity. The luc2P (1-hour half-life) gene responds more rapidly than luc2 (3-hour half-life) with moderate signal intensity, and the luc2CP (0.4-hour half-life) responds more quickly with the lowest signal intensity. The pGL4.23[luc2/minP], pGL4.24[luc2P/minP] and pGL4.25[luc2CP/minP] Vectors come without a selectable marker. To speed research, several pre-designed response element vectors based on the pGL4.27 Vector are available. Some of these also are available stable cell lines (GloResponse Cell Lines).