pGL4.75[hRluc/CMV] Vector, 20 µg, Promega
Catalog # PAE6931
Supplier: Promega Corporation
CAS Number:
Specifications
- Size:20 μg
- Environmentally Preferable:
- Cat. No.:PAE6931
- Storage temperature:–30...–10 °C
- Supplier no.:E6931
Specifications
About this item
pGL4.73[hRluc/SV40], pGL4.74[hRluc/TK] and pGL4.75[hRluc/CMV] Vectors are used as the control reporter for dual-luciferase assays. Select the appropriate promoter or test all three in your experimental system.
- Vectors Co-transfected in Dual-Reporter Assays as the Control Vector
- Co-transfect into cells where firefly luciferase is the experimental reporter gene
- CMV, SV40 and TK promoter options offer different expression levels suitable for most dual-reporter assays
The Promoter-driven Renilla control vectors are commonly co-transfected with experimental firefly luciferase vectors for use in the Dual-Luciferase or Dual-Glo Reporter Assay Systems. The control Renilla vectors should give an almost invariant level of activity, while the experimental firefly vector varies with treatment.