About this item
Highly purified, recombinant Taq DNA Polymerase cloned from thermophilic eubacteria, Thermus aquaticus.
- Ultrapure cloned Taq DNA polymerase
- Virtually eliminates background artifacts
- Minimizes smearing in long PCR
- Most thermostable Taq DNA polymerase
Taq2000 DNA Polymerase is a highly purified, recombinant Taq DNA polymerase cloned from the thermophilic eubacteria, Thermus aquaticus.