Order Entry
United States
Orders LinkContactUsLinkComponent
pRL-Null Vector, 20 µg, Promega

pRL-Null Vector, 20 µg, Promega

20 μg, –30...–10 °C
Catalog # PAE2271
Supplier:   Promega Corporation
Supplier #:  E2271
pRL-Null Vector, 20 µg, Promega
Catalog # PAE2271
CAS Number:  
pRL-Null Vector, 20 µg, Promega
pRL-Null Vector, 20 µg, Promega

20 μg, –30...–10 °C

CAS Number:  
Price
$427.65
Log in for availability
Each (20µG)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The pRL Vectors are wildtype Renilla luciferase control reporter vectors, which provide constitutive expression of Renilla luciferase and can be used in combination with a firefly luciferase vector to cotransfect mammalian cells.

  • Four Vectors that Contain Wild-Type Renilla Luciferase for Normalization in Reporter Assays
  • A T7 promoter is located immediately upstream of Rluc for in vitro synthesis of Renilla luciferase
  • The SV40 late poly(A) signal sequence is positioned downstream of Rluc to provide efficient transcription termination and mRNA polyadenylation
  • A prokaryotic origin of replication and β-lactamase gene allow selected propagation of the pRL vectors in E. coli host strains

The pRL Vectors are wildtype Renilla luciferase (Rluc) control reporter vectors. The pRL Vectors, which provide constitutive expression of Renilla luciferase, can be used in combination with a firefly luciferase vector to cotransfect mammalian cells. Expression of Renilla luciferase provides an internal control value to which expression of the experimental firefly luciferase reporter gene may be normalized. The pRL Vectors contain the cDNA encoding Renilla luciferase (Rluc) cloned from the anthozoan coelenterate Renilla reniformis (sea pansy). Four different promoter configurations are available. The HSV-thymidine kinase promoter (pRL-TK) is relatively weak and may be particularly useful in providing neutral constitutive expression of the Renilla luciferase control reporter. The early SV40 enhancer/promoter region (pRL-SV40) and the CMV immediate early enhancer/promoter region (pRL-CMV) typically provide high-level transcription and, therefore, may be less suitable for co-reporter applications involving experimental vectors with robust regulatory elements. In general, we recommend validating the performance of specific co-reporter combinations in the desired target cells. In addition to the modified Rluc reporter gene, all pRL Vectors are isolated from a dam-/dcm- E. coli K host strain, allowing digestion with restriction enzymes that are sensitive to dam and dcm methylation.

 

Specifications

Specifications

  • Size:
    20 μg
  • Storage temperature:
    –30...–10 °C

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.