Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ALE-0540
ALE-0540 ≥98% (by TLC)
ALE-0540 ≥98% (by TLC)
76203-384
:
Avantor
:
ALE-0540 ≥98% (by TLC)
76203-384
:
Avantor
:
Size:
100 mg
Cat. No.:
76203-384
:
:
301.254 g/mol
:
Refrigerator
:
234779-34-1
:
C₁₄H₁₁N₃O₅