Order Entry
United States
ContactUsLinkComponent
pGL3-Control Vector, 20 µg, Promega
pGL3-Control Vector, 20 µg, Promega
Catalog # PAE1741
Supplier:  Avantor
CAS Number:  
undefined
pGL3-Control Vector, 20 µg, Promega
Catalog # PAE1741
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAE1741
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    E1741

Specifications

About this item

Four firefly luciferase reporter vectors.

  • Four Reporter Vectors That Contain luc+, a Modified Firefly Luciferase Gene
  • Unique NcoI site located at 5´ end of luc+ gene will create fusions with reporter gene
  • SmaI site in the MCS means blunt-ended inserts can be ligated into the MCS and cut on either side by other restriction endonucleases
  • XbaI site just downstream of luc+ gene facilitates insertions into the 3´ untranslated region of mRNA or subcloning of the luciferase gene

The pGL3 Luciferase Reporter Vectors provide a basis for the quantitative analysis of factors that potentially regulate mammalian gene expression. These may be cis- or trans-acting factors. The backbone of the pGL2 Luciferase Reporter Vectors was redesigned for the pGL3 Vectors for increased expression, with a modified coding region for firefly (Photinus pyralis) luciferase that has been optimized for monitoring transcriptional activity in transfected eukaryotic cells. The assay of this genetic reporter is rapid, sensitive and quantitative. In addition, the Luciferase Reporter Vectors contain numerous features aiding in the structural characterization of the putative regulatory sequences under investigation. For the most advanced reporter vectors and widest selection of features, please see the pGL4 series of vectors.