Specifications
- Concentration:500 µg/ml in 10 mM Tris-HCl (pH 7.5), 1 mM EDTA
- Description:kB DNA Ladder
- Size:250 µg
- Environmentally Preferable:
- Cat. No.:ST201115
- Supplier no.:201115
Specifications
About this item
The supercoiled DNA ladder contains fragments from 2 to 10 kB at 1 kB increments and an additional band at 12 kB.
- Fragments are exact multiples or fractions of 1000 bp
- Includes useful reference point for determining location
- Economical
- Easy radiolabeling of ladder
- Known mass amount of each fragment for DNA quantitation
The kB DNA ladder contains fragments from 1 to 10 kB at 1 kB increments, as well as bands at 250 bp, 500 bp, 750 bp, 1500 bp and 12 kB.