Anti-Ki-67 Mouse Monoclonal Antibody
76203-746
- Antibody Type:Primary
- Antigen Name:Ki-67
- Antigen Symbol:Ki-67
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:1 ml
- Form:Liquid
- Antigen Synonyms:MKI67
- Storage Buffer:In Tris buffer, pH 7.5, containing 1% BSA and <0.1% sodium azide.
- Storage Temperature:+4°C
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Recombinant Ki-67.
- Purification:Affinity-purified.
- Cat. No.:76203-746
Recommended Applications: IHC
Type: Primary
Antigen: Ki-67
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human