T7 RNA Polymerase (HC), Promega
PAP4074
:
- Description:T7 RNA Polymerase (HC), 10,000u
- Size:10,000u
- Storage Conditions:-30°C to -10°C
- Enzyme Name:T7 RNA Polymerase
- Concentration:80 units/µL
- Tested Applications:Labeling in vitro, Transcription in vitro
- Cat. No.:PAP4074
- Supplier no.:P4074
DNA-dependent RNA polymerase for in vitro transcription.
- DNA-Dependent RNA Polymerase Specific for T7 Promoter Sequences
- >90% pure as determined by SDS polyacrylamide gel electrophoresis
- Free of detectable levels of contaminating RNase and DNase activity (
- Incorporates 32P, 33P, 3H and 35S nucleoside triphosphates
T7 RNA Polymerase is a DNA-dependent RNA polymerase that exhibits extremely high specificity for its cognate promoter sequences. Only T7 DNA or DNA cloned downstream from a T7 promoter can serve as a template for T7 RNA Polymerase-directed RNA synthesis.