Specifications
- Description:SEV Plungers, 50/pk
- Cat. No.:PAAS5201
- Supplier no.:AS5201
Specifications
About this item
Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration
- Standard Elution Volume (SEV) is 200 to 400 µl
- Low Elution Volume (LEV) is 30 to 100 µl
- Plungers and Elution Tubes for the Maxwell(R) 16 Instruments only (AS2000 and AS3000)
Maxwell 16 and Maxwell(R) 16 Forensic Instrument Accessories: Plungers, Elution Tubes, Hardware Kits, Cartridge Racks, Magnetic Elution Rack, etc.
Storage temperature: 15 to 30°C