Sca I, Restriction Enzymes, Promega
Catalog # PAR6211
Supplier: Avantor
CAS Number:
Specifications
- Description:Sca I
- Size:1000 U
- Storage Conditions:–30...–10 °C
- Enzyme Name:Sca I
- Buffer activity:% Activity (37°C) in 4 CORE® Buffers A: < 10% – B: 100%** – C: 50…75% – D: 75…100%; MULTI-CORE™ Buffer 10…25%
- Enzyme assay temperature:37°C
- Cat. No.:PAR6211
- Supplier no.:R6211
Specifications
About this item
AGT/ACT TCA\TGA
- Capable of digesting DNA in 15 minutes or less
- Available at high concentration, containing 5,000 units of ScaI at a concentration of 40-80u/microl