rUTP, 100 mM, 400 µl, Promega
Catalog # PAE6021
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:rUTP, 100mM, 400microl
- Size:400 μl
- Storage Conditions:-30°C to -10°C
- Cat. No.:PAE6021
- Supplier no.:E6021
Specifications
About this item
High-quality rNTPs. Provided at concentrations of 10mM or 100mM in water at neutral pH.
- High-Quality Ribonucleoside Triphosphates (rNTPs)
- 400µl rNTP supplied at 10mM or 100mM in water at neutral pH
- Qualified for use with the Riboprobe(R) and HeLaScribe(R) Systems
- Purity verified by HPLC
Ribonucleotide Triphosphates (rNTPs) are provided in individual tubes and qualified for use with the Riboprobe and HeLaScribe Systems. The rNTPs are supplied in nuclease-free water. Purity is verified by HPLC analysis.