RNAgents® Denaturing Solution, Promega
Catalog # PAZ5651
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:RNAgents® Denaturing Solution
- Size:120 ml
- Cat. No.:PAZ5651
- Supplier no.:Z5651
Specifications
About this item
RNAgents Denaturing Solution lyses cells or tissue under conditions that rapidly inhibit ribonucleases.
- Lyses Cells or Tissue Under Conditions that Rapidly Inhibit Ribonucleases
- Uses two potent inhibitors of RNase, guanidine thiocyanate and β-mercaptoethanol
- Designed to be used with acidic phenol:chloroform and alcohol (isopropanol) for purification of total RNA
- Fits RNAgents® Total RNA Isolation System (PAZ5110)
RNAgents Denaturing Solution lyses cells or tissue under conditions that rapidly inhibit ribonucleases, using two potent inhibitors of RNase, guanidine thiocyanate and beta-mercaptoethanol. The RNAgents Denaturing Solution is designed to be used in concert with acidic phenol:chloroform and alcohol (isopropanol) for purification of total RNA.