Order Entry
United States
ContactUsLinkComponent
RiboMAX Large Scale RNA Production System-SP6, 1 system, Promega
RiboMAX Large Scale RNA Production System-SP6, 1 system, Promega
Catalog # PAP1280
CAS Number:  
undefined
RiboMAX Large Scale RNA Production System-SP6, 1 system, Promega
Catalog # PAP1280
Supplier Number:  P1280
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    RiboMAX Large Scale RNA Production System-SP6, 1 system
  • Size:
    1 system
  • Cat. No.:
    PAP1280
  • Supplier no.:
    P1280

Specifications

About this item

Designed for large-scale RNA production, the RiboMAX Systems consistently produce 2-5mg RNA from a 1ml reaction volume.

  • Capped or Uncapped RNA Transcripts for in vitro or in vivo Translation
  • Produces 2-5mg/ml RNA in a 1ml reaction
  • Available for use with SP6 or T7 RNA polymerase
  • Scalable to meet varying RNA production requirements

The RiboMAX Large Scale RNA Production Systems consistently produce 2-5mg/ml of RNA in a 1ml reaction, about 10- to 20-fold more RNA than is produced with the standard Riboprobe System transcription reaction. The RiboMAX System reactions differ from those of the Riboprobe Systems in three primary ways: a HEPES (pH 7.5) buffer is used rather than a Tris-HCl (pH 7.9) buffer; rNTP and magnesium concentrations are elevated at levels appropriate for either SP6 or T7 RNA polymerase; and inorganic pyrophosphatase is included in the reaction. RNAs synthesized with the RiboMAX System perform better for in vitro translation in rabbit reticulocyte translation systems than RNA synthesized by standard methods. The reduction of components inhibitory to translation may be advantageous for other applications requiring biologically active RNA. Because the RiboMAX Systems produce large quantities of RNA, these systems are not recommended for the generation of high-specific-activity RNA probes. Note: Use of the RiboMAX System for production of capped transcripts requires separate purchase of the Ribo m7G Cap Analog (Cat. P1711).

Caution: Store at –20°C