Order Entry
United States
ContactUsLinkComponent
ReliaPrep RNA Tissue Miniprep System, Promega
ReliaPrep RNA Tissue Miniprep System, Promega
Certificates
undefined
ReliaPrep RNA Tissue Miniprep System, Promega
Certificates

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Storage Conditions:
    15°C to 30°C
  • Description:
    ReliaPrep RNA Tissue Miniprep System

Specifications

Related Information

Frequently Bought Together