Order Entry
United States
ContactUsLinkComponent
 
rCTP, rATP, rUTP, rGTP, 100 mM each, 4×400 microl, Promega
  PAE6000
 :  
undefined
rCTP, rATP, rUTP, rGTP, 100 mM each, 4×400 microl, Promega
  PAE6000
 :  E6000
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    rCTP, rATP, rUTP, rGTP, 100mM each, 4 x 400microl
  • Size:
    4 x 400 μl
  • Storage Conditions:
    Store HeLaScribe Nuclear Extract and Positive Control DNA at –70°C. Store other components at –20°C.
  • Cat. No.:
    PAE6000
  • Supplier no.:
    E6000

 

 

High-quality rNTPs. Set of four provided at concentrations of 10mM or 100mM in water at neutral pH.

  • High-Quality Ribonucleoside Triphosphates (rNTPs)
  • Provided at 10mM (4 x 0.5ml) or 100mM (4 x 400µl) in water at neutral pH
  • Qualified for use with the Riboprobe(R) and HeLaScribe(R) Systems
  • Purity verified by HPLC

Set of four rNTPs (rATP, rUTP, rCTP, rGTP) provided in individual tubes and qualified for use with the Riboprobe and HeLaScribe Systems. The rNTPs are supplied in nuclease-free water. Purity is verified by HPLC analysis.