Order Entry
United States
ContactUsLinkComponent
Rabbit Reticulocyte Lysate System, Nuclease Treated, 30 reactions, Promega
Rabbit Reticulocyte Lysate System, Nuclease Treated, 30 reactions, Promega
Catalog # PAL4960
CAS Number:  
undefined
Rabbit Reticulocyte Lysate System, Nuclease Treated, 30 reactions, Promega
Catalog # PAL4960
Supplier Number:  L4960
CAS Number:  

Specifications

  • Description:
    Rabbit Reticulocyte Lysate System, Nuclease Treated, 30 reactions
  • Size:
    30 reactions
  • Storage Temperature:
    LESS THAN -65°C
  • Cat. No.:
    PAL4960
  • Supplier no.:
    L4960

Specifications

About this item

Rabbit Reticulocyte Lysate Translation Systems are used in identification of mRNA species, characterization of their protein products and investigation of transcriptional and translational control.

  • In Vitro Translation of mRNA and Characterization of mRNA Products
  • Optimized for translation and ready to use
  • Contains an energy-regenerating system, a mix of tRNAs, hemin, potassium chloride and magnesium acetate

Rabbit Reticulocyte Lysate Translation Systems are utilized in the identification of mRNA species, the characterization of their protein products and the investigation of transcriptional and translational control. Rabbit Reticulocyte Lysate is prepared from New Zealand white rabbits using a standard protocol that ensures reliable and consistent reticulocyte production in each lot. After the reticulocytes are lysed, the extract is treated with micrococcal nuclease to destroy endogenous mRNA and thus reduce background translation to a minimum. The lysate contains the cellular components necessary for protein synthesis (tRNA, ribosomes, amino acids, initiation, elongation and termination factors).