Order Entry
United States
ContactUsLinkComponent
Proteasome-Glo Chymotrypsin-Like Cell-Based Assay, 2×50 ml, Promega
Proteasome-Glo Chymotrypsin-Like Cell-Based Assay, 2×50 ml, Promega
Catalog # PAG8662
CAS Number:  
undefined
Proteasome-Glo Chymotrypsin-Like Cell-Based Assay, 2×50 ml, Promega
Catalog # PAG8662
Supplier Number:  G8662
CAS Number:  

Specifications

  • Description:
    Proteasome-Glo Chymotrypsin-Like Cell-Based Assay, 2 x 50ml
  • Size:
    2 x 50ml
  • Storage Conditions:
    Store the Proteasome–Glo Assay components at –20°C
  • Storage Temperature:
    -30°C to -10°C
  • Cat. No.:
    PAG8662
  • Supplier no.:
    G8662

Specifications

About this item

The Proteasome-Glo Cell-Based Assays are homogeneous, luminescent assays that measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome complex in cultured cells.

  • Homogeneous, Luminescent Assays that Measure Protease Activities in Cultured Cells
  • Simple "add-mix-measure" protocol
  • Chymotrypsin-Like, Trypsin-Like and Caspase-Like activity data directly from cellular environment
  • Results within 10-30 minutes after reagent addition

The Proteasome-Glo Cell-Based Assays are homogeneous, luminescent assays that individually measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome complex in cultured cells. The 26S proteasome is a 2.5MDa multiprotein complex found in all eukaryotic cells. Proteasome-Glo Cell-Based Assays provide luminogenic proteasome substrates in buffers optimized for cell permeabilization, proteasome activity and luciferase activity. Addition of the Proteasome-Glo Cell-Based Reagent in an add-mix-measure format results in proteasome cleavage of the substrate and rapid generation of a luminescent signal produced by the luciferase reaction. The three luminogenic substrates used to monitor specific protease activities include: Suc-LLVY-aminoluciferin for chymotrypsin-like, Z-LRR-aminoluciferin for trypsin-like, and Z-nLPnLD-aminoluciferin for caspase-like activity. Each luminogenic substrate is added to a buffer system optimized for its specific proteasome activity and luciferase activity. The reagents are added to cells in culture, and the proteasome cleaves the substrates, releasing luciferin, which is consumed by luciferase, producing glow-type luminescence correlating to enzyme activity or inhibition. The Proteasome-Glo Cell-Based 3-Substrate System consists of three homogeneous bioluminescent assays that measure the three proteolytic activities associated with the proteasome in a cell-based format (each of these three assays also is available separately). The Proteasome-Glo 3-Substrate System consists of three homogeneous bioluminescent assays in a purified enzyme-based format (each of these three assays also is available separately).