Order Entry
United States
ContactUsLinkComponent
PIP2:3PS Lipid Kinase Substrate, 0.25mg, 0.25ml, Promega
PIP2:3PS Lipid Kinase Substrate, 0.25mg, 0.25ml, Promega
Catalog # PAV1701
CAS Number:  
undefined
PIP2:3PS Lipid Kinase Substrate, 0.25mg, 0.25ml, Promega
Catalog # PAV1701
Supplier Number:  V1701
CAS Number:  

Specifications

  • Description:
    PIP2:3PS Lipid Kinase Substrate, 0.25mg, 0.25ml
  • Size:
    0.25ml
  • Storage Temperature:
    LESS THAN -65°C
  • Cat. No.:
    PAV1701
  • Supplier no.:
    V1701

Specifications

About this item

The lipid kinase substrates PIP2:3PS and PI:3PS are standalone substrates that are also part of the PI3K-Glo kinase profiling system.

  • Phosphoinositide-3 KInase (PI3K) Substrates
  • Use in screening assays with purified PI3K Class I lipid kinases and ADP-Glo™ Kinase Assay
  • PIP2:3PS supplied at 0.25mg in 0.25ml
  • PI:3PS supplied at 0.5mg in 0.5ml

The lipid kinase substrates PIP2:3PS and PI:3PS are standalone substrates that are part of the PI3K-Glo kinase profiling system. Promega lipid kinase enzymes, substrates and detection systems provide a complete set of reagents for performing phosphoinositide lipid kinase (PIK) reactions using a luminescent ADP-detection platform, the ADP-Glo Kinase Assay. The lipid kinase reaction is performed by incubating lipid substrate (PI:3PS or PIP2:3PS) with a recombinant enzyme and ATP, and the kinase activity is measured using the ADP-Glo Kinase Assay.