ContactUsLinkComponent
pGEM Express Positive Control Template, 10 µg, Promega
  PAP2561
 :  
undefined
pGEM Express Positive Control Template, 10 µg, Promega
  PAP2561
 :  P2561
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    pGEM Express Positive Control Template, 10microg
  • Size:
    10 μg
  • Cat. No.:
    PAP2561
  • Supplier no.:
    P2561
The pGEM Express Positive Control Template is used to monitor in vitro transcription reactions when using the Riboprobe Systems.
  • Used with Riboprobe(R) Systems to Monitor in vitro Transcription Reactions
  • Can be used with SP6, T7 or T3 RNA polymerase
  • SP6 RNA polymerase produces transcripts of 1,787 and 2,566 bases; T7 RNA polymerase produces transcripts of 1,065 and 2,346 bases; T3 RNA Polymerase produces transcripts of 250 and 1,525 bases
The pGEM Express Positive Control Template is created by linearizing a vector with the restriction enzyme ScaI. The Positive Control Template may be used to monitor in vitro transcription reactions when using the Riboprobe Systems.
 : Store at –20°C