About this item
PCR Nucleotide Mix is a premixed solution containing the sodium salts of dATP, dCTP, dGTP and dTTP, each at a concentration of 10mM or 25mM in water.
- cGMP-Grade Solution of dNTPs Pretested in PCR
- cGMP-manufactured for lot-to-lot product consistency
- Equimolar amounts of each dNTP in water
- Two concentrations available: 10mM and 25mM
High-quality deoxynucleotide triphosphates (dNTPs) are critical for PCR success. PCR Nucleotide Mix is a cGMP-grade premixed solution containing the sodium salts of dATP, dCTP, dGTP and dTTP. There are two formulations available: A premixed solution containing each nucleotide at a concentration of 10mM in water at pH 7.5; the total concentration of nucleotides is 40mM. A premixed solution containing each nucleotide at a concentration of 25mM in water at pH 7.5; the total concentration of nucleotides is 100mM. These PCR nucleotide mixes are ready to use and optimized for use in PCR.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Storage Conditions:-30°C to -10°C