Order Entry
United States
ContactUsLinkComponent
BenchTop PCR Markers, 300 µl, Promega
BenchTop PCR Markers, 300 µl, Promega
Catalog # PAG7531
CAS Number:  
undefined
BenchTop PCR Markers, 300 µl, Promega
Catalog # PAG7531
Supplier Number:  G7531
CAS Number:  

Specifications

  • Description:
    BenchTop PCR Markers, 300microl
  • Size:
    300 μl
  • Storage Temperature:
    -30°C to 30°C
  • Cat. No.:
    PAG7531
  • Supplier no.:
    G7531

Specifications

About this item

50-1,000bp; 6 bands. Can be stored at room temperature. Contains dye marker, ready to load on agarose and polyacrylamide gels.

  • Six Bands of Equal Intensity: 50, 150, 300, 500, 750, and 1,000bp
  • Store at room temperature (22-25°C)
  • Premixed with loading buffer; ready to load onto agarose gels
  • Load 6microl/lane for agarose gels

The BenchTop DNA Markers offer the convenience of storage at room temperature (22-25°C) as well as the capability of direct loading onto agarose gels. The BenchTop DNA Markers are supplied in a stabilizing solution of 1X Blue/Orange Loading Dye, which circumvents any requirements for further manipulation. Six bands of equal intensity of 50, 150, 300, 500, 750, and 1,000bp. The BenchTop PCR Markers may be run on polyacrylamide gels with less loading volume; however, additional bands may be visible compared to those visible on agarose gels. Load 6microl/lane for agarose gels.