Order Entry
United States
ContactUsLinkComponent
BenchTop 1 kb DNA Ladder, 600 µl, Promega
BenchTop 1 kb DNA Ladder, 600 µl, Promega
Catalog # PAG7541
CAS Number:  
undefined
BenchTop 1 kb DNA Ladder, 600 µl, Promega
Catalog # PAG7541
Supplier Number:  G7541
CAS Number:  

Specifications

  • Description:
    BenchTop 1kb DNA Ladder, 600microl
  • Size:
    600 μl
  • Storage Temperature:
    -30°C to 30°C
  • Cat. No.:
    PAG7541
  • Supplier no.:
    G7541

Specifications

About this item

250-10,000bp; 13 bands. Can be stored at room temperature. Contains dye marker, ready to load.

  • Thirteen Blunt-Ended Fragments of 250-10,000bp
  • Store at room temperature (22-25°C)
  • Premixed with loading buffer; ready to load onto agarose gels
  • Load 6microl/lane

The BenchTop DNA Markers offer the convenience of storage at room temperature (22-25°C) as well as the capability of direct loading onto agarose gels. The BenchTop DNA Markers are supplied in a stabilizing solution of 1X Blue/Orange Loading Dye, which circumvents any requirements for further manipulation. BenchTop 1kb DNA Ladder: Thirteen blunt-ended fragments with sizes ranging from 250bp to 10,000bp. The 1,000bp and 3,000bp fragments have increased intensity relative to the other bands on ethidium bromide-stained agarose gels for easy identification. All other fragments are of equal intensity. The ladder is dephosphorylated and ready for 5´ end-labeling with radioisotopes using T4 Polynucleotide Kinase, allowing visualization by autoradiography. Load 6microl/lane.