NADPH Regeneration System, Promega
Catalog # PAV9510
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:NADPH Regeneration System, 1,000 assays
- Size:1,000 assays
- Storage Temperature:-30°C to -10°C
- Cat. No.:PAV9510
- Supplier no.:V9510
Specifications
About this item
The NADPH Regeneration System is an enzyme substrate combination that reduces NADP+ to NADPH.
- A component of the P450-Glo CYP450 Screening Systems
- Available for purchase separately
The NADPH Regeneration System is an enzyme substrate combination that reduces NADP+ to NADPH. The system consists of two reagents, Solution A and Solution B. Solution A contains the substrate NADP+ and glucose-6-phosphate and is supplied as a 20X concentrate. Solution B contains the enzyme glucose-6-phosphate dehydrogenase at a 100X concentration. The two solutions are combined before use, and the NADPH generated serves as the source of electrons for the P450 oxidative reactions.