ContactUsLinkComponent
MS Compatible Yeast Protein Extract, Digest
MS Compatible Yeast Protein Extract, Digest
  PAV7461
 :  
undefined
MS Compatible Yeast Protein Extract, Digest
  PAV7461
 :  V7461
 :  
  • Size:
    100 μg
  • Storage Conditions:
    –30...–10 °C
  • Protein/Peptide Name:
    MS Compatible Yeast Protein Extract, Digest
  • Cat. No.:
    PAV7461
  • Supplier no.:
    V7461
Mass spectrometry-compatible yeast and human protein extracts are designed specifically for mass spectrometry applications. The extracts are predigested and cleaned up by solid-phase extraction for immediate use in liquid chromatography/mass spec analysis
  • Protein Extracts for Easier Mass Spec Instrument Monitoring
  • Choose human or yeast, digested or intact extract
  • Monitor LC and MS instrument performance
  • Use for method development and optimization
The mass spectrometry-compatible yeast and human protein extracts are designed specifically for mass spectrometry applications (i.e., instrument monitoring). The extracts are predigested and cleaned up by solid-phase extraction for immediate use in liquid chromatography/mass spectrometry (LC/MS) analysis. Both the yeast and human extracts also are available in an intact undigested form to provide a test material for optimizing mass spec protein sample preparation. The yeast extracts are beneficial for users who prefer working with a relatively compact and well studied proteome, whereas the human extract provides opportunity for working with a complex proteome having a large dynamic range. Consistent extract protein composition is ensured by tight control over cell culture conditions and manufacturing process. Lot-to-lot consistency of extracts is monitored by various protein and peptide qualitative and quantitation methods, including LC/MS and amino acid analysis. Our manufacturing process assures compatibility with reverse phase liquid chromatography and mass spectrometry by monitoring nonspecific protein fragmentation, nonbiological post-translational modifications and, for digested extracts, minimal undigested peptides.