ContactUsLinkComponent
M-MLV Reverse Transcriptase, RNase H Minus, Promega
M-MLV Reverse Transcriptase, RNase H Minus, Promega
Catalog # PAM5301
CAS Number:  
undefined
M-MLV Reverse Transcriptase, RNase H Minus, Promega
Catalog # PAM5301
Supplier Number:  M5301
CAS Number:  
Specifications
  • Description:
    M-MLV Reverse Transcriptase, RNase H Minus, 10,000u
  • Size:
    10,000u
  • Cat. No.:
    PAM5301
  • Supplier no.:
    M5301
Specifications
About this item
Deletion mutation of M-MLV RT resulting in no RNase H activity. Suitable for rare or long transcripts.
  • Helpful for Preparing Long cDNAs and Libraries with a High Percentage of Full-Length cDNA
  • Supplied at a concentration of 100-200u/microl
Moloney Murine Leukemia Virus Reverse Transcriptase, RNase H Minus (M-MLV RT [H-]), is an RNA-dependent DNA polymerase that can be used in cDNA synthesis with long messenger RNA templates (>5kb). This form of M-MLV Reverse Transcriptase has been genetically altered to remove the associated RNase H activity. Although many researchers are successful in using M-MLV RT (H+) for analytical and some preparative cDNA applications, reverse transcriptases lacking RNase H activity provide another option to prepare long cDNAs and libraries containing a high percentage of full-length cDNA.
Caution: Storage temperature -30°C to -10°C