Order Entry
United States
ContactUsLinkComponent
MethylEdge Bisulfite Conversion System, 50 reactions, Promega
MethylEdge Bisulfite Conversion System, 50 reactions, Promega
Catalog # PAN1301
CAS Number:  
undefined
MethylEdge Bisulfite Conversion System, 50 reactions, Promega
Catalog # PAN1301
Supplier Number:  N1301
CAS Number:  

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Methylated DNA
  • Sample type:
    DNA preps
  • Target molecule:
    DNA
  • Description:
    MethylEdge Bisulfite Conversion System, 50 reactions
  • Size:
    50 reactions
  • Storage Conditions:
    15°C to 30°C
  • Description:
    MethylEdge Bisulfite Conversion System
  • Cat. No.:
    PAN1301
  • Supplier no.:
    N1301

Specifications

About this item

The MethylEdge Bisulfite Conversion System provides a rapid, efficient method to perform bisulfite conversion with minimal DNA fragmentation in less than two hours.

  • Rapid Bisulfite Conversion with Minimal DNA Fragmentation
  • High-efficiency DNA conversion from as little as 100pg input DNA
  • Rapid protocol saves time compared to other conversion systems
  • Robust conversion of DNA with reduced DNA fragmentation

The MethylEdge Bisulfite Conversion System provides a rapid, efficient method to perform bisulfite conversion with minimal DNA fragmentation in less than two hours. The rapid protocol and complete conversion mean that you can produce completely converted DNA ready for your choice downstream assays with minimal preparation and hands-on time. Coupled with purification chemistries and flexible, robust amplification technologies for detection, the MethylEdge Bisulfite Conversion System delivers high-quality, intact converted DNA for your experiments.