MagneSil ONE, Fixed Yield Blood Genomic System, 1 x 96 preps, Promega
Catalog # PAMD1370
Supplier: Promega Corporation
CAS Number:
Specifications
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:Blood
- Target molecule:DNA
- Description:MagneSil ONE, Fixed Yield Blood Genomic System, 1 x 96 preps
- Size:1 x 96 preps
- Storage Conditions:15°C to 30°C
- Description:MagneSil ONE, Fixed Yield Blood Genomic System
- Cat. No.:PAMD1370
- Supplier no.:MD1370
Specifications
About this item
The MagneSil ONE, Fixed Yield Blood Genomic System purifies 1ug of DNA (+/- 50%) from 60ul of anti-coagulated whole blood.
- Automated Extraction of 1µg DNA from 60µl Blood
- Process 96 samples in 1 hour with no hands-on time following robot setup
- DNA suitable for use in PCR, multiplex PCR and SNP genotyping applications
- Validated automated methods available
The MagneSil ONE, Fixed Yield Blood Genomic System purifies 1ug of DNA (+/- 50%) from 60ul of anti-coagulated whole blood. Purification of a fixed yield of DNA eliminates the need to quantitate and normalize concentrations postpurification. The highly pure DNA isolated is suitable for use in PCR, multiplex PCR and SNP genotyping applications. Walkaway automation is available on the Beckman Coulter Biomek FX in a 96-well format. Process 96 samples in about 1 hour with no hands-on time following robot setup.