Order Entry
United States
ContactUsLinkComponent
MagneSil Blood Genomic, Max Yield System, 1 x 96 preps, Promega
MagneSil Blood Genomic, Max Yield System, 1 x 96 preps, Promega
Catalog # PAMD1360
CAS Number:  
undefined
MagneSil Blood Genomic, Max Yield System, 1 x 96 preps, Promega
Catalog # PAMD1360
Supplier Number:  MD1360
CAS Number:  

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
  • Target molecule:
    DNA
  • Description:
    MagneSil Blood Genomic, Max Yield System, 1 x 96 preps
  • Size:
    1 x 96 preps
  • Storage Conditions:
    15°C to 30°C
  • Description:
    MagneSil Blood Genomic, Max Yield System
  • Cat. No.:
    PAMD1360
  • Supplier no.:
    MD1360

Specifications

About this item

MagneSil Blood Genomic, Max Yield System provides automated high-throughput DNA purification on the Beckman Coulter Biomek FX using MagneSil Paramagnetic Particle technology.

  • Walkaway Automation of Genomic DNA Extraction
  • Purifies DNA from 96 samples of anti-coagulated whole blood in 1 1/2 hours
  • Average yield of 96 samples from normal healthy adults is ≥4microg
  • Validated automated methods available

The MagneSil Blood Genomic, Max Yield System provides automated high-throughput DNA purification on the Beckman Coulter Biomek FX using MagneSil Paramagnetic Particle technology. DNA from 96 samples of anti-coagulated human whole blood is purified in about 1 1/2 hours with no hands-on time once the robot protocol is initiated. Studies on DNA recovery and purity and PCR results show no cross-contamination between samples in adjacent wells. Purified DNA is qualified for single-locus simple PCR as well as more demanding applications such as multiplex PCR (e.g., PowerPlex 16 System [Cat.# DC6531], Y Chromosome Deletion Detection System [Cat.# MD1531]) and SNP genotyping.