Order Entry
United States
ContactUsLinkComponent
Luciferase SP6 Control DNA, Promega
Luciferase SP6 Control DNA, Promega
Catalog # PAL4741
CAS Number:  
undefined
Luciferase SP6 Control DNA, Promega
Catalog # PAL4741
Supplier Number:  L4741
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Luciferase SP6 Control DNA
  • Size:
    20 μg
  • Storage Temperature:
    -30°C to -10°C
  • Cat. No.:
    PAL4741
  • Supplier no.:
    L4741

Specifications

About this item

Luciferase SP6 and T7 Control DNAs are used with the TNT Quick Coupled and TNT Coupled Transcription/Translation Systems. The Control DNAs contain the gene for luciferase under transcriptional control of a phage RNA polymerase promoter.

  • Functional Controls for the TnT(R) Quick Coupled and TnT(R) Coupled Transcription/Translation Systems
  • Contains luciferase gene under transcriptional control of an SP6 or T7 phage RNA polymerase promoter
  • Includes a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene
  • Easy to monitor control reactions with a luciferase assay

The Luciferase SP6 and T7 Control DNAs are used as functional controls in the TNT Quick Coupled and TNT Coupled Transcription/Translation Systems. The Control DNAs contain the gene for luciferase under transcriptional control of a phage RNA polymerase promoter. All constructs carry a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene. Control reactions are monitored easily by the production of luminescence, which is generated from full-length luciferase and the addition of necessary components. Luciferase Control DNAs are supplied as 0.5mg/ml solutions in TE buffer.