ContactUsLinkComponent
Luciferase Control RNA, Promega
  PAL4561
 :  
undefined
Luciferase Control RNA, Promega
  PAL4561
 :  L4561
 :  
  • Description:
    Luciferase Control RNA
  • Size:
    20 μg
  • Environmentally Preferable:
  • Storage Temperature:
    LESS THAN -65°C
  • Cat. No.:
    PAL4561
  • Supplier no.:
    L4561
Luciferase Control RNA is an uncapped in vitro-transcribed RNA containing a 30-base poly(A) tail that produces functional luciferase when translated.
  • Unique Functional Control to Monitor In Vitro Translation Reactions
  • Control reactions are easily monitored using a luciferase assay
  • Safe, non-radioactive format to monitor control activity
  • Supplied at a concentration of 1mg/ml
Luciferase Control RNA is a unique functional control for in vitro translation reactions. Luciferase Control RNA is an uncapped in vitro-transcribed RNA containing a 30-base poly(A) tail that produces functional luciferase when translated. Control reactions are monitored easily by a luciferase assay for the production of luminescence generated from the full-length luciferase.