Order Entry
United States
ContactUsLinkComponent
 
Kinase Selectivity Profiling System: Other/CK-1 + ADP-Glo Assay, 8 × 50 reactions, Promega
Catalog # PAV6919
CAS Number:  
undefined
Kinase Selectivity Profiling System: Other/CK-1 + ADP-Glo Assay, 8 × 50 reactions, Promega
Catalog # PAV6919
Supplier Number:  V6919
CAS Number:  

Specifications

  • Description:
    Kinase Selectivity Profiling System: Other/CK-1 + ADP-Glo Assay, 8 x 50 reactions
  • Size:
    8 x 50 reactions
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
  • Cat. No.:
    PAV6919
  • Supplier no.:
    V6919

Specifications

About this item

Easy-to-use kits for performing kinase selectivity profiling that rely on the ADP-Glo Kinase Assay technology. Each system includes kinase and substrate pairs in an easy-to-use 8-tube strip format optimized for fast and simple kinase profiling reactions.

  • Selectivity Screening of Library Compounds for Effect on a Panel of Kinases
  • Kinase and substrate pairs organized in an easy-to-use 8-tube strip format
  • Simple reaction assembly and automation-friendly protocol
  • Streamilines profiling of hits and leads for potency against families of related kinases

The Kinase Selectivity Profiling Systems are easy-to-use kits for performing kinase selectivity profiling that rely on the ADP-Glo Kinase Assay technology. Each system includes kinase and substrate pairs organized in an easy-to-use 8-tube strip format optimized for fast and simple kinase profiling reactions. Kinase Selectivity Profiling Systems offer kinases grouped either in single kinase family strips or as a general panel of kinases representative of the human kinome for a broad kinase profile. Each profiling system contains the reagents needed to complete a profile for a compound, including kinase reaction buffer, eight kinases in each multiwell strip and eight corresponding substrates and cofactors in another multiwell strip. If we do not offer a prebuilt kinase selectivity profiling system that meets your experimental needs, you can design your own custom system using the custom kinase selectivity profiling system tool (www.promega.com/products/cell-signaling/kinase-assays/customize/).