Order Entry
United States
ContactUsLinkComponent
Interface Tubing and Valves for Auto Injector System, 1 each, Promega
Interface Tubing and Valves for Auto Injector System, 1 each, Promega
Catalog # PAE2211
Interface Tubing and Valves for Auto Injector System, 1 each, Promega
Catalog # PAE2211
Supplier Number:  E2211

Specifications

  • Description:
    Interface Tubing and Valves for Auto Injector System
  • Size:
    1 each
  • Storage Temperature:
    15°C to 30°C
  • Cat. No.:
    PAE2211
  • Supplier no.:
    E2211

Specifications

About this item

These accessories are available for the now-discontinued 20/20 luminometer.

  • For Use With TD-20/20 Instrument
  • Accessories only for discontinued Turner BioSystems Luminometer Model TD-20/20
  • Not for Turner BioSystems 20/20n Single-Tube Luminometer or Promega GloMax(R) 20/20 Luminometer
  • Power Supply now discontinued

These accessories are only for the now-discontinued Turner BioSystems Luminometer Model TD-20/20. They are not for the Turner BioSystems 20/20n Single-Tube Luminometer or Promega GloMax 20/20 Luminometer. Turner BioSystems and Promega have teamed up to develop the next-generation luminometers for single-tube and 96-well plate analysis. For more information, see the GloMax Luminometer catalog pages or contact Technical Support.