ContactUsLinkComponent
HaloTag Cloning Starter System, 1 each, Promega
HaloTag Cloning Starter System, 1 each, Promega
Catalog # PAG6050
CAS Number:  
HaloTag Cloning Starter System, 1 each, Promega
Catalog # PAG6050
Supplier Number:  G6050
CAS Number:  
Specifications
  • Description:
    HaloTag Cloning Starter System
  • Size:
    1 each
  • Cat. No.:
    PAG6050
  • Supplier no.:
    G6050
Specifications
About this item
HaloTag Cloning Starter Kit: designed to use Flexi Cloning System to clone protein coding sequences into mammalian HaloTag expression vectors; clone and transfer gene sequences into C- or N-terminus of HaloTag Flexi CMV Deletion Vectors.
  • HaloTag Cloning Starter Kit: designed to use Flexi Cloning System to clone protein coding sequences into mammalian HaloTag expression vectors; clone and transfer gene sequences into C- or N-terminus of HaloTag Flexi CMV Deletion Vectors.
The HaloTag Cloning Starter Kit is designed to utilize the Flexi Cloning System to clone protein coding sequences into mammalian HaloTag expression vectors. The system contains reagents required to clone and transfer gene sequences into the C- or N-terminus of a variety of HaloTag Flexi CMV Deletion Vectors. This series of vectors containing deletions of human cytomegalovirus (CMV) immediate-early enhancer/promoter were created to provide different constitutive expression levels of HaloTag 7 fusion proteins, providing greater control over the level of protein expression. This allows expression of enough fusion protein to image or capture but not so much to disturb normal function of the protein partner fused to the HaloTag 7 protein.