About this item
Taq DNA Polymerase. Provided with Green and Colorless Reaction Buffers. Green Buffer allows direct gel analysis after amplification. Enhanced performance over standard Taq DNA Polymerase.
- Taq DNA Polymerase with Buffers for Enhanced PCR Amplification
- Direct-to-gel convenience
- Can be used with your current PCR cycling conditions
- Supplied at a concentration of 5u/microl
Experience improved PCR performance with GoTaq DNA Polymerase products. GoTaq DNA Polymerase is a proprietary formulation of Taq DNA polymerase that gives robust amplification equal to and in some cases superior to that of standard Taq DNA polymerase. The 5X GoTaq Green and Colorless Reaction Buffers supplied with GoTaq DNA Polymerase contain MgCl2 at a concentration of 7.5mM for a final concentration of 1.5mM in the 1X reaction. The 5X Green and 5X Colorless Reaction Buffers supplied with GoTaq enzymes allow you to go directly from thermal cycler to gel analysis. These buffers contain a compound that increases sample density, so that samples sink easily into wells of an agarose gel. The green buffer also contains two dyes (yellow and blue) that separate to allow easy monitoring during electrophoresis. The blue dye comigrates at the same rate as 3-5kb DNA fragments in a 1% agarose gel. The yellow dye migrates ahead of primers (<50bp). Use the green reaction buffer for direct-to-gel analysis after amplification and the colorless reaction buffer for post-amplification analysis by fluorescence or absorbance without prior DNA purification.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Environmentally Preferable: