ENLITEN ATP Assay System, Promega
Catalog # PAFF2000
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:ENLITEN ATP Assay System, 100 assays
- Size:100 assays
- Storage Conditions:Store at –20°C unopened. See product insert for individual component storage conditions before and after opening.
- Storage Temperature:-30°C to -10°C
- Cat. No.:PAFF2000
- No. of assays/kit:100
- Supplier no.:FF2000
Specifications
About this item
The ENLITEN ATP Assay System is used to measure ATP levels for the indirect detection of biocontamination.
- Allows Indirect Detection of Biocontamination
The ENLITEN ATP Assay System can be used to measure ATP levels for the indirect detection of biocontamination on food processing surfaces, in cosmetics and beverages or to assay for enzymes that degrade ATP and to quantitate ATP in biological fluids.