About this item
Endo H is cloned from S. plicatus and overexpressed in E. coli.
- Characterize Protein Glycosylation
- Monitor protein trafficking
- Characterize glycan location and structure
- Does not cleave complex glycans
- Supplied at a concentration of 500u/microl
Endoglycosidase H (Endo H) is a recombinant glycosidase cloned from Streptomyces plicatus and overexpressed in E. coli. Endo H cleaves the chitobiose core of high mannose and a limited number of hybrid oligosaccharides from N-linked glycoproteins. It does not cleave complex glycans. Enzymatic cleavage is between the two N-acetylglucosamine residues in the diacetylchitobiose core of the oligosaccharide, leaving one N-acetylglucosamine residue on the asparagine. This is in contrast to PNGase F, which cleaves all asparagine-linked oligosaccharides.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Storage Conditions:–30...–10 °C
- Enzyme Name:Endo H