Order Entry
United States
ContactUsLinkComponent
dCTP, 200 µmol, Promega
dCTP, 200 µmol, Promega
Catalog # PAU1222
CAS Number:  
undefined
dCTP, 200 µmol, Promega
Catalog # PAU1222
Supplier Number:  U1222
CAS Number:  

Specifications

  • Description:
    dCTP, 200micromol
  • Size:
    200 μmol
  • Storage Conditions:
    -30°C to -10°C
  • Concentration:
    100 mM
  • Cat. No.:
    PAU1222
  • Supplier no.:
    U1222

Specifications

About this item

High-quality dNTPs. Provided at a concentration of 100mM in water at pH 7.5.

  • High-Quality Nucleotides for Your Workflow
  • Greater than 99% triphosphate content
  • 100mM in water at pH 7.5

High-quality deoxynucleotide triphosphates (dNTPs) are critical for the success of many key procedures such as cDNA synthesis, sequencing and labeling. Promega dNTPs have greater than 99% triphosphate content and are provided at a concentration of 100mM in water at pH 7.5.