CellTiter-Glo® Luminescent Cell Viability Assay, Promega
About this item
The most sensitive, fastest, homogeneous cell viability assay. Based on luminescent detection of ATP.
- A Luminescent Assay to Determine the Number of Viable Cells in Culture
- A homogeneous "add-mix-measure" format reduces the number of steps
- Higher level of detection requires fewer cells per assay
- Process plates consecutively with a very stable luminescent signal
The CellTiter-Glo Luminescent Cell Viability Assay is a homogeneous method of determining the number of viable cells in culture based on quantitation of the ATP present, an indicator of metabolically active cells. The CellTiter-Glo Assay is designed for use with multiwell formats, making it ideal for automated high-throughput screening (HTS), cell proliferation and cytotoxicity assays. The homogeneous assay procedure involves adding the single reagent (CellTiter-Glo Reagent) directly to cells cultured in serum-supplemented medium. Cell washing, removal of medium and multiple pipetting steps are not required. The system detects as few as 15 cells/well in a 384-well format in 10 minutes after adding reagent and mixing. The homogeneous add-mix-measure format results in cell lysis and generation of a luminescent signal proportional to the amount of ATP present. The amount of ATP is directly proportional to the number of cells present in culture. The CellTiter-Glo Assay generates a glow-type luminescent signal, which has a half-life generally greater than five hours, depending on cell type and medium used. The extended half-life eliminates the need to use reagent injectors and provides flexibility for continuous or batch mode processing of multiple plates. The unique homogeneous format avoids errors that may be introduced by other ATP measurement methods that require multiple steps.
4 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The most sensitive, fastest, homogeneous cell viability assay. Based on luminescent detection of ATP.
- A Luminescent Assay to Determine the Number of Viable Cells in Culture
- A homogeneous "add-mix-measure" format reduces the number of steps
- Higher level of detection requires fewer cells per assay
- Process plates consecutively with a very stable luminescent signal
The CellTiter-Glo Luminescent Cell Viability Assay is a homogeneous method of determining the number of viable cells in culture based on quantitation of the ATP present, an indicator of metabolically active cells. The CellTiter-Glo Assay is designed for use with multiwell formats, making it ideal for automated high-throughput screening (HTS), cell proliferation and cytotoxicity assays. The homogeneous assay procedure involves adding the single reagent (CellTiter-Glo Reagent) directly to cells cultured in serum-supplemented medium. Cell washing, removal of medium and multiple pipetting steps are not required. The system detects as few as 15 cells/well in a 384-well format in 10 minutes after adding reagent and mixing. The homogeneous add-mix-measure format results in cell lysis and generation of a luminescent signal proportional to the amount of ATP present. The amount of ATP is directly proportional to the number of cells present in culture. The CellTiter-Glo Assay generates a glow-type luminescent signal, which has a half-life generally greater than five hours, depending on cell type and medium used. The extended half-life eliminates the need to use reagent injectors and provides flexibility for continuous or batch mode processing of multiple plates. The unique homogeneous format avoids errors that may be introduced by other ATP measurement methods that require multiple steps.
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- PAG7573
- PAG7570
- PAG7571
- PAG7572
- Storage Temperature:
- -30°C to -10°C
- -30°C to -10°C
- -30°C to -10°C
- -30°C to -10°C
Specifications
Product Family Options
Product Information
- SizeVolumeNo. of assays/kitAvailabilityPrice
- PAG7573CellTiter-Glo Luminescent Cell Viability AssayCellTiter-Glo Luminescent Cell Viability Assay10 x 100 ml10× 100 ml10000 - 40000Log in for availabilityEach$5,439.18Supplier Number: G7573Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.PAG7573Storage Temperature-30°C to -10°C - PAG7570CellTiter-Glo Luminescent Cell Viability AssayCellTiter-Glo Luminescent Cell Viability Assay10 ml10 ml100 - 400Log in for availabilityEach$161.76Supplier Number: G7570Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.PAG7570Storage Temperature-30°C to -10°C - PAG7571CellTiter-Glo Luminescent Cell Viability AssayCellTiter-Glo Luminescent Cell Viability Assay10 x 10 ml10× 10 ml1000 - 4000Log in for availabilityEach$835.09Supplier Number: G7571Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.PAG7571Storage Temperature-30°C to -10°C - PAG7572CellTiter-Glo Luminescent Cell Viability AssayCellTiter-Glo Luminescent Cell Viability Assay100 ml100 ml1000 - 4000Log in for availabilityEach$660.18Supplier Number: G7572Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.PAG7572Storage Temperature-30°C to -10°C