ContactUsLinkComponent
Passive Lysis 5X Buffer, Promega
Passive Lysis 5X Buffer, Promega
Catalog # PAE1941
CAS Number:  
undefined
Passive Lysis 5X Buffer, Promega
Catalog # PAE1941
Supplier Number:  E1941
CAS Number:  
Specifications
  • Description:
    Passive Lysis 5X Buffer
  • Size:
    30 mL
  • Cat. No.:
    PAE1941
  • Supplier no.:
    E1941
Specifications
About this item
Promotes rapid lysis of cultured mammalian cells without the need to scrape adherent cells or freeze-thaw. Lytic reagent of choice when processing cells for quantitation of firefly and Renilla luciferase activities using the Promega Dual-Luciferase® Reporter Assay (#PAE1910).
  • Rapid Lysis of Cultured Mammalian Cells
  • No need for scraping adherent cells or performing freeze-thaw cycles
  • Minimal coelenterazine autoluminescence
Caution: For Research Use Only. Not for Use in Diagnostic Procedures.