Single Stage High Purity Brass Regulators, PerkinElmer
97013-482
- Description:Single stage high purity brass regulators for air
- Cat. No.:97013-482
- Delivery pressure gauge:0 - 4000
- Delivery range:10 - 200
- Supplier no.:00230090
High purity brass regulators are constructed of high purity brass barstock and have stainless steel diaphragms and metal-to-metal seals. They are suitable for use with high purity (>99.995% pure) non-corrosive gases.
- Regulators terminate in ¹/₄"
- NPT swagelock fitting
Ideally suited for chromatographic carrier gas applications including FID, TCD,ECD, HID, and non-corrosive gas mixtures for analytical instrumentation.
: Supplied with adapter.