Dulbecco's Phosphate-Buffered Saline (DPBS), Himedia®
Supplier: HiMedia
Certificates
About this item
Dulbecco's phosphate buffered saline is used for tissue disaggregation and monolayer dispersal.The presence of calcium and magnesium ions hinder the trypsin activity.
- Clear, colorless solution
- pH: 7.20 to 7.80
The solution is used to maintain the medium within physiological pH range and intracellular and extra cellular osmotic balance. Modified with a carbohydrate, such as glucose serves as an energy source for cell metabolism.
Specifications
- Shelf Life:24 months
Specifications
Frequently Bought Together